Famciclovir
Famciclovir
Famciclovir
Thermo Scientific Chemicals

Famciclovir

Have Questions?
Número de catálogoCantidad
4631700505 g
Número de catálogo 463170050
Precio (EUR)
159,80
Online Exclusive
193,00
Ahorro 33,20 (17%)
Each
Disponibilidad estimada  Pendientes  
Añadir al carro de la compra
Cantidad:
5 g
Pedido a granel o personalizado
Precio (EUR)
159,80
Online Exclusive
193,00
Ahorro 33,20 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS104227-87-4
IUPAC Name2-[(acetyloxy)methyl]-4-(2-amino-9H-purin-9-yl)butyl acetate
Molecular FormulaC14H19N5O4
InChI KeyGGXKWVWZWMLJEH-UHFFFAOYSA-N
SMILESCC(=O)OCC(CCN1C=NC2=CN=C(N)N=C12)COC(C)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point103°C to 107°C
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0389269Certificate of Analysis02 jun 2025463170010, 463170050
A0389269Certificate of Analysis01 dic 2023463170010, 463170050
K05W008Certificate of Analysis22 dic 2021463170010, 463170050
Se muestran 3 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection