Clorhidrato de cinacalcet
Thermo Scientific Chemicals

Clorhidrato de cinacalcet

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 466010010
Precio (EUR)
131,75
Precio exclusivo en nuestra web
159,00
Ahorro 27,25 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
1 g
Pedido a granel o personalizado
Precio (EUR)
131,75
Precio exclusivo en nuestra web
159,00
Ahorro 27,25 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS364782-34-3
IUPAC Name[(1R)-1-(naphthalen-1-yl)ethyl]({3-[3-(trifluoromethyl)phenyl]propyl})amine hydrochloride
Molecular FormulaC22H23ClF3N
InChI KeyQANQWUQOEJZMLL-PKLMIRHRSA-N
SMILESCl.C[C@@H](NCCCC1=CC=CC(=C1)C(F)(F)F)C1=C2C=CC=CC2=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Specific optical rotation(C=0.5 in Methanol)
Appearance (Form)powder
Appearance (Color)White to beige
Specific optical rotation-32.0° to -24.0° (20°C, 589 nm)
Solubility(2 mg/ml in DMSO) Clear
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0418085Certificate of Analysis17 may 2025466010050, 466010010
A0418085Certificate of Analysis27 oct 2022466010050, 466010010
Se muestran 2 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection