L-Erythrulose
Thermo Scientific Chemicals

L-Erythrulose

CAS: 533-50-6 | C4H8O4, C4H8O4 | 120.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 467300250
Price (EUR)
80,50
Online Exclusive
94,70
Save 14,20 (15%)
Each
-
Add to cart
Quantity:
25 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
80,50
Online Exclusive
94,70
Save 14,20 (15%)
Each
Add to cart
Chemical Identifiers
CAS533-50-6
IUPAC Name(3S)-1,3,4-trihydroxybutan-2-one
Molecular FormulaC4H8O4
InChI KeyUQPHVQVXLPRNCX-YEQUQEJONA-N
SMILESOC[C@H](O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<24.0 %
Infrared spectrumConforms
Appearance (Color)Yellow to green to brown
HPLC>=76.0 %
Sulfated ash=<4.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472338Certificate of AnalysisMay 28, 2025467300250, 467301000
A0419586Certificate of AnalysisJan 25, 2025467300250, 467301000
A0419586Certificate of AnalysisAug 25, 2023467300250, 467301000
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection