Thermo Scientific Chemicals

Dichlorobis(triphenylphosphine)nickel(II), 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): 013930.18
50 g, Each
Thermo Scientific Chemicals

Dichlorobis(triphenylphosphine)nickel(II), 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): 013930.18
50 g, Each
数量
製品番号(カタログ番号): 013930.18
または、製品番号013930-18

化学物質識別子

CAS
14264-16-5
IUPAC Name
dichloronickelbis(ylium); bis(triphenylphosphane)
Molecular Formula
C36H30Cl2NiP2
InChI Key
VHQPRJDILRYZBG-UHFFFAOYSA-L
SMILES
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
> 97.0%
Appearance (Color)
Green to black
Form
crystalline powder

概要

Dichlorobis(triphenylphosphine)nickel(II) is used as a catalyst for cross-coupling of Grignard reagents, hydrosilylations, hydrogenation and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text