Germanium(II) iodide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Germanium(II) iodide, 99.99% (metals basis), Thermo Scientific Chemicals
製品番号(カタログ番号) 014013.03
または、製品番号014013-03
価格(JPY)/ Each
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13573-08-5
Molecular FormulaGeH6I2
InChI KeyPUZROLYCAWSHNL-UHFFFAOYSA-N
SMILES[GeH4].I.I
Molecular Weight (g/mol)332.49
Total Metal Impurities0.01% max.
Germanium(II) iodide acts as a reagent that forms stable ionic adducts with carbenes. It is also used in the preparation of allylgermanes by reacting with allylstannanes. Further, it reacts with allyl bromide in the presence of zinc iodide to get allylgermanium(IV), which on reacts with carbonyl compounds to give the corresponding homoallylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only