Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
数量:
100 g
5 g
25 g
製品番号(カタログ番号) 038325.22
または、製品番号038325-22
価格(JPY)/ Each
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
Surface Area900-1100 m²/g
Assay from Supplier's CofAMetal content (Pt): 9.5-10.5%
Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jeweler. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-
RUO – Research Use Only