Thermo Scientific Chemicals

3,4,9,10-Perylenetetracarboxylic diimide, Thermo Scientific Chemicals

製品番号(カタログ番号): 044098.06
5 g, Each
Thermo Scientific Chemicals

3,4,9,10-Perylenetetracarboxylic diimide, Thermo Scientific Chemicals

製品番号(カタログ番号): 044098.06
5 g, Each
数量
製品番号(カタログ番号): 044098.06
または、製品番号044098-06

化学物質識別子

CAS
81-33-4
IUPAC Name
7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular Formula
C24H10N2O4
InChI Key
KJOLVZJFMDVPGB-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=CC=C3C4=CC=C5C(=O)NC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
Appearance (Color)
Purple red
Form
Powder
Assay from Supplier's CofA
≥94%

概要

Used in chemical research. PTCDI is used extensively as an industrial pigment. It is used as tunable laser dye, light-harvesting material, transistor, molecular switched, solar cell, and optoelectronic device. Planar perylene- and naphthalene-based diimide linkers can be employed to tether the Watson- Crick and the Hoogsteen strands of a DNA triplex, thus providing conjugates capable of targeting singlestranded nucleic acids with the formation of hairpin triplexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text