Thermo Scientific Chemicals

Lead(II) iodide, 98.5%, Thermo Scientific Chemicals

製品番号(カタログ番号): 057103.30
250 g, Each
Thermo Scientific Chemicals

Lead(II) iodide, 98.5%, Thermo Scientific Chemicals

製品番号(カタログ番号): 057103.30
250 g, Each
数量
製品番号(カタログ番号): 057103.30
または、製品番号057103-30

化学物質識別子

CAS
10101-63-0
IUPAC Name
λ²-lead(2+) diiodide
Molecular Formula
I2Pb
InChI Key
RQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES
[I-].[I-].[Pb++]
Assay (unspecified)
98.5% min

概要

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text