1x1 image pixel for data collection
Potassium permanganate, 99+%, ACS reagent
Potassium permanganate, 99+%, ACS reagent
Potassium permanganate, 99+%, ACS reagent
Potassium permanganate, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium permanganate, 99+%, ACS reagent

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol
製品番号(カタログ番号) 424175000
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7722-64-7
IUPAC Namepotassium manganesoylolate
Molecular FormulaKMnO4
InChI KeyVZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES[K+].[O-][Mn](=O)(=O)=O
さらに表示
Appearance (Form)Glistening crystalline powder
Titration with KMnO4>=99.0 %
Insoluble matter=<0.2 %
Chloride and Chlorate (as Cl)=<0.005 %
Sulfate (SO4)=<0.02 %
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0473924Certificate of Analysis2025年6月27日424175000, 424170025
A0467522Certificate of Analysis2025年1月25日424175000, 424170025
A0468580Certificate of Analysis2025年1月25日424175000, 424170025
A0466334Certificate of Analysis2024年9月25日424175000, 424170025
A0461538Certificate of Analysis2024年3月31日424175000, 424170025
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection