2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 38614-36-7 | C4H7BrMg | 159.31 g/mol
数量:
100 mL
800 mL
パッケージ:
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 435561000
価格(JPY)
-
見積もりを依頼する
数量:
100 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS38614-36-7
IUPAC Namebromo(2-methylprop-1-en-1-yl)magnesium
Molecular FormulaC4H7BrMg
InChI KeyBKVIQODYSREJQH-UHFFFAOYSA-M
SMILESCC(C)=C[Mg]Br
さらに表示
Appearance (Color)Yellow to green to brown
Appearance (Form)Clear to hazy solution
AppearanceThis material can vary from light to very dark in
Appearanceappearance. These differences in color will have
Appearanceno effect on use.
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。