Fexofenadine hydrochloride
Product Image
Thermo Scientific Chemicals

Fexofenadine hydrochloride

CAS: 153439-40-8 | C32H40ClNO4 | 538.13 g/mol
製品番号(カタログ番号) 464970010
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS153439-40-8
IUPAC Namehydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
Molecular FormulaC32H40ClNO4
InChI KeyRRJFVPUCXDGFJB-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to almost white
Infrared spectrumConforms
Loss on drying=<1 %
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection