Thermo Scientific Chemicals

3-Nitro-L-tyrosine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11018.14
25g, Each
Thermo Scientific Chemicals

3-Nitro-L-tyrosine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11018.14
25g, Each
数量
製品番号(カタログ番号): A11018.14
または、製品番号A11018-14

化学物質識別子

CAS
621-44-3
IUPAC Name
(2S)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
Molecular Formula
C9H10N2O5
InChI Key
FBTSQILOGYXGMD-MDOHGIEYNA-N
SMILES
N[C@@H](CC1=CC=C(O)C(=C1)[N+]([O-])=O)C(O)=O
Appearance (Color)
Yellow
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥97.5% (HPLC, U.S. sourced material)
Identification (FTIR)
Conforms (UK sourced material)

概要

3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.

Solubility
Soluble in water (partly miscible), 1N hydrochloric acid (50 mg/ml), acidic aqueous solution , ethanol, DMSO, DMF and PBS (pH 7.2) (∼2 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text