Thermo Scientific Chemicals

Terephthaloyl chloride, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11224.30
250 g, Each
Thermo Scientific Chemicals

Terephthaloyl chloride, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11224.30
250 g, Each
数量
製品番号(カタログ番号): A11224.30
または、製品番号A11224-30

化学物質識別子

CAS
100-20-9
IUPAC Name
benzene-1,4-dicarbonyl dichloride
Molecular Formula
C8H4Cl2O2
InChI Key
LXEJRKJRKIFVNY-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
White
Form
crystalline flakes or powder
Assay (GC)
> 98.5%
Assay (Titration ex Chloride)
> 98.5%

概要

Terephthaloyl chloride acts as a monomer with p-phenylenediamine used in the preparation of polymer viz. poly paraphenylene terephthalamide. It is an active component in performance polymers and aramid fibers, which gives flame resistance, chemical resistance, temperature stability, light weight, and very high strength. It is also used as an effective water scavenger, utilized to stabilize isocyanates and urethane prepolymers. Further, it is used in the preparation of liquid crystalline thermosets by thermal cyclotrimerization of dicyanate compounds of ring substituted bis(4-hydroxyphenyl) terepthalates. In addition to this, it is involved in the condensation reaction with difunctional alfa, μ-diaminopolystyrene to get chain-extended polystyrene containing amide bonds along the polymer backbone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terephthaloyl chloride acts as a monomer with p-phenylenediamine used in the preparation of polymer viz. poly paraphenylene terephthalamide. It is an active component in performance polymers and aramid fibers, which gives flame resistance, chemical resistance, temperature stability, light weight, and very high strength. It is also used as an effective water scavenger, utilized to stabilize isocyanates and urethane prepolymers. Further, it is used in the preparation of liquid crystalline thermosets by thermal cyclotrimerization of dicyanate compounds of ring substituted bis(4-hydroxyphenyl) terepthalates. In addition to this, it is involved in the condensation reaction with difunctional alfa, µ-diaminopolystyrene to get chain-extended polystyrene containing amide bonds along the polymer backbone.

Solubility
Soluble in ethanol, ether and other common organic solvents.

Notes
Incompatible with alcohols, oxidizing agents, water and strong bases.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text