Thermo Scientific Chemicals

2-Amino-4,6-dichloropyrimidine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11330.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-4,6-dichloropyrimidine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11330.06
5 g, Each
数量
製品番号(カタログ番号): A11330.06
または、製品番号A11330-06

化学物質識別子

CAS
56-05-3
IUPAC Name
4,6-dichloropyrimidin-2-amine
Molecular Formula
C4H3Cl2N3
InChI Key
JPZOAVGMSDSWSW-UHFFFAOYSA-N
SMILES
NC1=NC(Cl)=CC(Cl)=N1
Appearance (Color)
Pale cream
Melting Point (clear melt)
218.0-225.0?C
Assay (GC)
≥97.5%
Form
Powder

概要

Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.

Solubility
Insoluble in water, soluble in acetone, two monomer (VCM) and thermal toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text