Octafluorotoluene, 98+%, Thermo Scientific Chemicals
Octafluorotoluene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Octafluorotoluene, 98+%, Thermo Scientific Chemicals

数量:
25 g
5 g
100 g
製品番号(カタログ番号) A11453.14
または、製品番号A11453-14
価格(JPY)/ Each
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS434-64-0
IUPAC Name1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
Molecular FormulaC7F8
InChI KeyUSPWUOFNOTUBAD-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
さらに表示
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.3655-1.3695 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only