Thermo Scientific Chemicals

3-Butyn-1-ol, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11477.06
5 g, Each
Thermo Scientific Chemicals

3-Butyn-1-ol, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11477.06
5 g, Each
数量
製品番号(カタログ番号): A11477.06
または、製品番号A11477-06

化学物質識別子

CAS
927-74-2
IUPAC Name
but-3-yn-1-ol
Molecular Formula
C4H6O
InChI Key
OTJZCIYGRUNXTP-UHFFFAOYSA-N
SMILES
OCCC#C
Refractive Index
1.4385-1.4425 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid

概要

3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.

Solubility
Miscible with alcohols and organic solvents. Immiscible with aliphatic hydrocarbon solvents.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides and copper.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text