Thermo Scientific Chemicals

Diethyl acetylenedicarboxylate, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11719.14
25 g, Each
Thermo Scientific Chemicals

Diethyl acetylenedicarboxylate, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号): A11719.14
25 g, Each
数量
製品番号(カタログ番号): A11719.14
または、製品番号A11719-14

化学物質識別子

CAS
762-21-0
IUPAC Name
1,4-diethyl but-2-ynedioate
Molecular Formula
C8H10O4
InChI Key
STRNXFOUBFLVIN-UHFFFAOYSA-N
SMILES
CCOC(=O)C#CC(=O)OCC
Refractive Index
1.4405-1.4465 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%

概要

Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

Solubility
Soluble in ethanol, ethyl ether, CCl4, Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with strong acids and strong bases.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text