Thermo Scientific Chemicals

2,4-Hexadienal, predominantly trans,trans, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13200.06
5 g, Each
Thermo Scientific Chemicals

2,4-Hexadienal, predominantly trans,trans, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13200.06
5 g, Each
数量
製品番号(カタログ番号): A13200.06
または、製品番号A13200-06

化学物質識別子

CAS
142-83-6
IUPAC Name
(2E,4E)-hexa-2,4-dienal
Molecular Formula
C6H8O
InChI Key
BATOPAZDIZEVQF-MQQKCMAXSA-N
SMILES
CC=CC=CC=O
Identification (FTIR)
Conforms
Appearance (Color)
Clear yellow
Free acid (titration)
≤2.5%
Refractive Index
1.5390-1.5450 @ 20?C
Assay (GC)
≥94.0%

概要

2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.

Solubility
It is Insoluble in water, but soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Store under inert gas.Take measures to prevent the build up of electrostatic charge. Incompatible with strong reducing agents, strong bases, strong oxidizing agents. It is Stable, Combustible.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text