Thermo Scientific Chemicals

Hexanoic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13789.0F
2500 mL, Each
Thermo Scientific Chemicals

Hexanoic acid, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13789.0F
2500 mL, Each
数量
製品番号(カタログ番号): A13789.0F
または、製品番号A13789-0F

化学物質識別子

CAS
142-62-1
IUPAC Name
hexanoic acid
Molecular Formula
C6H12O2
InChI Key
FUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILES
CCCCCC(O)=O
Assay (Silylated GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Liquid
Identification (FTIR)
Conforms

概要

Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

Solubility
Miscible with diethyl ether and ethanol. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and allyl alcohol.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text