Thermo Scientific Chemicals

1-Bromo-4-iodobenzene, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13859.18
50 g, Each
Thermo Scientific Chemicals

1-Bromo-4-iodobenzene, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13859.18
50 g, Each
数量
製品番号(カタログ番号): A13859.18
または、製品番号A13859-18

化学物質識別子

CAS
589-87-7
IUPAC Name
1-bromo-4-iodobenzene
Molecular Formula
C6H4BrI
InChI Key
UCCUXODGPMAHRL-UHFFFAOYSA-N
SMILES
BrC1=CC=C(I)C=C1
Appearance (Color)
White to cream or pink to brown
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Form
Crystalline powder
Melting Point (clear melt)
89-93?C

概要

It is mainly used as the OLED pharmaceutical intermediate, as reagent for in situ desilylation and coupling of silylated alkynes, as substrate in copper-free Sonogashira coupling in aqueous acetone. It is also used in synthesis of β,β,dibromostyrenes, as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used as the OLED pharmaceutical intermediate, as reagent for in situ desilylation and coupling of silylated alkynes, as substrate in copper-free Sonogashira coupling in aqueous acetone. It is also used in synthesis of β,β,dibromostyrenes, as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids).

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, light.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text