Thermo Scientific Chemicals

3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15338.03
1 g, Each
Thermo Scientific Chemicals

3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15338.03
1 g, Each
数量
製品番号(カタログ番号): A15338.03
または、製品番号A15338-03

化学物質識別子

CAS
26153-38-8
IUPAC Name
3,5-dihydroxybenzaldehyde
Molecular Formula
C7H6O3
InChI Key
HAQLHRYUDBKTJG-UHFFFAOYSA-N
SMILES
OC1=CC(C=O)=CC(O)=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%

概要

3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Air sensitive. Store in cool place. Incompatible with strong acids and strong bases.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text