Thermo Scientific Chemicals

Methyl o-toluate, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15695.14
25 g, Each
Thermo Scientific Chemicals

Methyl o-toluate, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15695.14
25 g, Each
数量
製品番号(カタログ番号): A15695.14
または、製品番号A15695-14

化学物質識別子

CAS
89-71-4
IUPAC Name
methyl 2-methylbenzoate
Molecular Formula
C9H10O2
InChI Key
WVWZECQNFWFVFW-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1C
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5175-1.5205 @ 20°C
Assay (GC)
≥98.5%
Form
Liquid

概要

It has been used in the preparation of 2-(azidomethyl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the preparation of 2-(azidomethyl)benzoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text