Thermo Scientific Chemicals

1,4-Benzodioxane, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A17558.18
50 g, Each
Thermo Scientific Chemicals

1,4-Benzodioxane, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A17558.18
50 g, Each
数量
製品番号(カタログ番号): A17558.18
または、製品番号A17558-18

化学物質識別子

CAS
493-09-4
IUPAC Name
2,3-dihydro-1,4-benzodioxine
Molecular Formula
C8H8O2
InChI Key
BNBQRQQYDMDJAH-UHFFFAOYSA-N
SMILES
C1COC2=CC=CC=C2O1
Form
Liquid
Refractive Index
1.5475-1.5515 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

概要

Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text