Thermo Scientific Chemicals

2-Chlorobutane, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): B21586.AD
50 mL, Each
Thermo Scientific Chemicals

2-Chlorobutane, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): B21586.AD
50 mL, Each
数量
製品番号(カタログ番号): B21586.AD
または、製品番号B21586-AD

化学物質識別子

CAS
78-86-4
IUPAC Name
2-chlorobutane
Molecular Formula
C4H9Cl
InChI Key
BSPCSKHALVHRSR-UHFFFAOYNA-N
SMILES
CCC(C)Cl
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.3945-1.3985 @ 20?C

概要

2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

Solubility
Immiscible with water.

Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text