Thermo Scientific Chemicals

4-Bromo-2,5-dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): B25500.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2,5-dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号): B25500.14
25 g, Each
数量
製品番号(カタログ番号): B25500.14
または、製品番号B25500-14

化学物質識別子

CAS
31558-41-5
IUPAC Name
4-bromo-2,5-dimethoxybenzaldehyde
Molecular Formula
C9H9BrO3
InChI Key
BIFWGDWGCZLCHF-UHFFFAOYSA-N
SMILES
COC1=CC(Br)=C(OC)C=C1C=O
Assay (GC)
>98.0%
Appearance (Color)
White to yellow
Form
Crystals or Crystalline powder
Free acid (titration)
<1.0%

概要

4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text