Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MAQLFLPLLA ALVLAQAPAA LADVLEGDSS EDRAFRVRIA GDAPLQGVLG GALTIPCHVH YLRPPPSRRA VLGSPRVKWT FLSRGREAEV LVARGVRVKV NEAYRFRVAL PAYPASLTDV SLALSELRPN DSGIYRCEVQ HGIDDSSDAV EVKVKGVVFL YREGSARYAF SFSGAQEACA RIGAHIATPE QLYAAYLGGY EQCDAGWLSD QTVRYPIQTP REACYGDMDG FPGVRNYGVV DPDDLYDVYC YAEDLNGELF LGDPPEKLTL EEARAYCQER GAEIATTGQL YAAWDGGLDH CSPGWLADGS VRYPIVTPSQ RCGGGLPGVK TLFLFPNQTG FPNKHSRFNV YCFRDSAQPS AIPEASNPAS NPASDGLEAI VTVTETLEEL QLPQEATESE SRGAIYSIPI MEDGGGGSST PEDPAEAPRT LLEFETQSMV PPTGFSEEEG KALEEEEKYE DEEEKEEEEE EEEVEDEALW AWPSELSSPG PEASLPTEPA AQEKSLSQAP ARAVLQPGAS PLPDGESEAS RPPRVHGPPT ETLPTPRERN LASPSPSTLV EAREVGEATG GPELSGVPRG ESEETGSSEG APSLLPATRA PEGTRELEAP SEDNSGRTAP AGTSVQAQPV LPTDSASRGG VAVVPASGDC VPSPCHNGGT CLEEEEGVRC LCLPGYGGDL CDVGLRFCNP GWDAFQGACY KHFSTRRSWE EAETQCRMYG AHLASISTPE EQDFINNRYR EYQWIGLNDR TIEGDFLWSD GVPLLYENWN PGQPDSYFLS GENCVVMVWH DQGQWSDVPC NYHLSYTCKM GLVSCGPPPE LPLAQVFGRP RLRYEVDTVL RYRCREGLAQ RNLPLIRCQE NGRWEAPQIS CVPRRPARAL HPEEDPEGRQ GRLLGRWKAL LIPPSSPMPG P
Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEHAB; Brain-enriched hyaluronan-binding protein; Brevican core protein; Chondroitin sulfate proteoglycan 7; chondroitin sulfate proteoglycan BEHAB
Gene Aliases: BCAN; BEHAB; CSPG7; UNQ2525/PRO6018
UniProt ID: (Human) Q96GW7
Entrez Gene ID: (Human) 63827
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support