Thermo Scientific Chemicals

6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H26794.06
5 g, Each
Thermo Scientific Chemicals

6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H26794.06
5 g, Each
数量
製品番号(カタログ番号): H26794.06
または、製品番号H26794-06

化学物質識別子

CAS
1122-71-0
IUPAC Name
(6-methylpyridin-2-yl)methanol
Molecular Formula
C7H9NO
InChI Key
JLVBSBMJQUMAMW-UHFFFAOYSA-N
SMILES
CC1=CC=CC(CO)=N1
Melting Point (clear melt)
28.0-34.0?C
Form
Fused solid
Appearance (Color)
White to pale cream
Assay (GC)
≥97.5%

概要

Use as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as primary and secondary intermediate.

Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text