Thermo Scientific Chemicals

4-Chloro-6,7-dimethoxyquinazoline, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H28405.14
25 g, Each
Thermo Scientific Chemicals

4-Chloro-6,7-dimethoxyquinazoline, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H28405.14
25 g, Each
数量
製品番号(カタログ番号): H28405.14
または、製品番号H28405-14

化学物質識別子

CAS
13790-39-1
IUPAC Name
4-chloro-6,7-dimethoxyquinazoline
Molecular Formula
C10H9ClN2O2
InChI Key
LLLHRNQLGUOJHP-UHFFFAOYSA-N
SMILES
COC1=C(OC)C=C2C(Cl)=NC=NC2=C1
Melting Point (clear melt)
182.0-188.0?C
Appearance (Color)
Pale yellow
Form
Powder
Assay (GC)
≥97.5%

概要

A useful synthetic intermediate in the preparation of epidermal growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate in the preparation of epidermal growth factors.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text