Thermo Scientific Chemicals

3-Chloro-4-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): H32038.03
1 g, Each
Thermo Scientific Chemicals

3-Chloro-4-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): H32038.03
1 g, Each
数量
製品番号(カタログ番号): H32038.03
または、製品番号H32038-03

化学物質識別子

CAS
115754-20-6
IUPAC Name
3-chloro-4-(trifluoromethyl)benzoic acid
Molecular Formula
C8H4ClF3O2
InChI Key
UDXPRKSPAZWHQN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(Cl)=C(C=C1)C(F)(F)F
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
155.0-164.0?C
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%

概要

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text