Thermo Scientific Chemicals

3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): H52069.03
1 g, Each
Thermo Scientific Chemicals

3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): H52069.03
1 g, Each
数量
製品番号(カタログ番号): H52069.03
または、製品番号H52069-03

概要

3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text