Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals

製品番号(カタログ番号): H54632.AE
100 mL, Each
Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals

製品番号(カタログ番号): H54632.AE
100 mL, Each
数量
製品番号(カタログ番号): H54632.AE
または、製品番号H54632-AE

概要

3,4-(Methylenedioxy)phenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agen
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text