Thermo Scientific Chemicals

Methyl 2-fluoro-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H61675.03
1 g, Each
Thermo Scientific Chemicals

Methyl 2-fluoro-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H61675.03
1 g, Each
数量
製品番号(カタログ番号): H61675.03
または、製品番号H61675-03

化学物質識別子

CAS
197507-22-5
IUPAC Name
methyl 2-fluoro-4-hydroxybenzoate
Molecular Formula
C8H7FO3
InChI Key
WYSPMXSNCAFCFV-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(F)C=C(O)C=C1
Assay (HPLC)
>97.5%

概要

Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text