Thermo Scientific Chemicals

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H62772.06
5 g, Each
Thermo Scientific Chemicals

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H62772.06
5 g, Each
数量
製品番号(カタログ番号): H62772.06
または、製品番号H62772-06

化学物質識別子

CAS
3350-13-8
IUPAC Name
2-{[(benzyloxy)carbonyl]amino}-4-{[(tert-butoxy)carbonyl]amino}butanoic acid; N-cyclohexylcyclohexanamine
Molecular Formula
C29H47N3O6
InChI Key
CYMIEBREXUYHGN-UHFFFAOYNA-N
SMILES
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Assay (HPLC)
>97.5%

概要

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text