Thermo Scientific Chemicals

2-Fluoro-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): H63235.06
5 g, Each
Thermo Scientific Chemicals

2-Fluoro-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): H63235.06
5 g, Each
数量
製品番号(カタログ番号): H63235.06
または、製品番号H63235-06

化学物質識別子

CAS
317-46-4
Assay (HPLC)
>96.0%

概要

2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text