Thermo Scientific Chemicals

2-(4-Bromophenyl)ethylamine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H63706.06
5 g, Each
Thermo Scientific Chemicals

2-(4-Bromophenyl)ethylamine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H63706.06
5 g, Each
数量
製品番号(カタログ番号): H63706.06
または、製品番号H63706-06

化学物質識別子

CAS
73918-56-6
IUPAC Name
2-(4-bromophenyl)ethan-1-aminium
Molecular Formula
C8H11BrN
InChI Key
ZSZCXAOQVBEPME-UHFFFAOYSA-O
SMILES
[NH3+]CCC1=CC=C(Br)C=C1
Assay (GC)
>97.5%

概要

4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text