Thermo Scientific Chemicals

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): H64329.03
1 g, Each
Thermo Scientific Chemicals

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): H64329.03
1 g, Each
数量
製品番号(カタログ番号): H64329.03
または、製品番号H64329-03

化学物質識別子

CAS
887707-25-7
Assay (HPLC)
>94.0%

概要

It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text