Thermo Scientific Chemicals

4-Bromo-3-chloro-2-fluoropyridine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H64362.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-3-chloro-2-fluoropyridine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): H64362.06
5 g, Each
数量
製品番号(カタログ番号): H64362.06
または、製品番号H64362-06

化学物質識別子

CAS
1017793-21-3
IUPAC Name
4-bromo-3-chloro-2-fluoropyridine
Molecular Formula
C5H2BrClFN
InChI Key
UMQBGWKWOLRROL-UHFFFAOYSA-N
SMILES
FC1=NC=CC(Br)=C1Cl
Assay (HPLC)
>97.5%

概要

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text