Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals

製品番号(カタログ番号): J63144.30
250 g, Each
Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals

製品番号(カタログ番号): J63144.30
250 g, Each
数量
製品番号(カタログ番号): J63144.30
または、製品番号J63144-30

化学物質識別子

CAS
115-69-5
IUPAC Name
2-amino-2-methylpropane-1,3-diol
Molecular Formula
C4H11NO2
InChI Key
UXFQFBNBSPQBJW-UHFFFAOYSA-N
SMILES
CC(N)(CO)CO
Assay (unspecified)
>99.0%

概要

A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

Solubility
Soluble in water and other organic solvents.

Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, aluminum, copper, brass and strong acids.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text