Yeast Lytic Enzyme, Arthrobacter luteus
Yeast Lytic Enzyme, Arthrobacter luteus
Thermo Scientific Chemicals
Yeast Lytic Enzyme, Arthrobacter luteus
Hydrolyzes linear glucose polymers with beta-1,3-linkages
数量:
250 mg
500 mg
1 g
製品番号(カタログ番号) J63195.MD
または、製品番号J63195-MD
価格(JPY)/ Each
-
見積もりを依頼する
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS37340-57-1
DNaseTrace level
Unit definitionOne unit of lytic acitvity is defined as amount which indicates 30% of decrease in absorbance at A800nm
FormLyophilized powder
RNaseTrace level
units/mgdwReport value
さらに表示
Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
2°C to 8°C