Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 99+%, Electrophoresis Grade

製品番号(カタログ番号): J66710.14
25 g, Each
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 99+%, Electrophoresis Grade

製品番号(カタログ番号): J66710.14
25 g, Each
数量
製品番号(カタログ番号): J66710.14
または、製品番号J66710-14

化学物質識別子

CAS
110-26-9
IUPAC Name
N-[(prop-2-enamido)methyl]prop-2-enamide
Molecular Formula
C7H10N2O2
InChI Key
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES
C=CC(=O)NCNC(=O)C=C
Assay (special)
Acrylic acid: ≤ 0.02%
Conductivity
≤ 10 µ mho/cm (2% in water)
pH
≥5.0 (2.5% aqueous solution)
Form
Crystalline powder
Protease activity
None detected

概要

N,N'-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.

Solubility
Soluble in water or methanol

Notes
Air & Light Sensitive. Store in the dark. Store under dry inert gas. Incompatible with light, heat, air and oxidizing agents. Store at 4°C.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text