Pasteurella multocida Toxin (PMT) Detection Kit
Pasteurella multocida Toxin (PMT) Detection Kit
Thermo Scientific™
Pasteurella multocida Toxin (PMT) Detection Kit
Rapidly detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs with this assay.
製品番号(カタログ番号)数量
K000921-9960 Tests (10 Plates)
製品番号(カタログ番号) K000921-9
価格(JPY)
-
見積もりを依頼する
数量:
960 Tests (10 Plates)
Ask our AI about this Product
Detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs1, with Thermo Scientific™ Oxoid™ Pasteurella multocida Toxin PMT Kit. The enzyme-linked immunosorbent assay is designed to deliver specific, sensitive, and fast detection of Pasteurella multocida toxin in porcine samples.
Progressive atrophic rhinitis is a disease of pigs, caused by infection with toxigenic P. multocida.1 The pathogenesis of the disease is linked to a single, well-characterized component: the Pasteurella multocida toxin (PMT). The clinical manifestations of the disease are shortening or distortion of the snout, sneezing, nasal discharge, and epitaxis. Reduced growth rate is seen in severe cases. Use ELISA Systems Pasteurella multocida Toxin PMT Detection Kit for the detection of Pasteurella multocida toxin in porcine samples.
  • Simple: Ready-to-use liquid reagents and concentrates make the assay easy to implement and run
  • Sensitive: Detect proteins at low levels for improved assurance of safety
  • Specific: Detect PMT in any type of sample, including extracts of primary mixed cultures obtained from, e.g., nasal swabs or washings2
Other names for the PMT include dermonecrotic toxin (DNT), heat labile toxin (HLT), turbinate atrophy toxin (TAT), and osteolytic toxin. For Veterinary Use Only. Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
General References:
  1. Pedersen KB, Nielsen JP, Foged NT, Elling F, Nielsen NC, Willeberg P. Atrophic rhinitis in pigs: proposal for a revised definition. Vet Rec 1988; 122: 190-1.
  2. Foged NT, Nielsen JP, Pedersen KB. Differentiation of toxigenic from non-toxigenic Pasteurella multocida by enzymelinked immunosorbent assay, Journal of Clinical Microbiology, 1988; 26: 1419-20.
仕様
概要Pasteurella multocida Toxin (PMT) Detection Kit
検出可能な分析物Pasteurella multocida toxin
イムノアッセイキットのフォーマットELISA (Standard)
キット内容Conjugate, Microtitration plate, Positive control antigen, Reconstitution buffer, Sealing tape, Stop solution, Substrate, Wash buffer concentrate, White strips
数量960 Tests (10 Plates)
保存要件2°C to 8°C
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
ドキュメントおよびダウンロード
証明書
Safety Data Sheets
こちらの製品には複数の製品番号が設定されています。