Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): L03540.22
100 g, Each
Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): L03540.22
100 g, Each
数量
製品番号(カタログ番号): L03540.22
または、製品番号L03540-22

化学物質識別子

CAS
140-28-3
IUPAC Name
benzyl[2-(benzylamino)ethyl]amine
Molecular Formula
C16H20N2
InChI Key
JUHORIMYRDESRB-UHFFFAOYSA-N
SMILES
C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0% (non-U.S. specification)
Comment
Purchased in U.S. and in other countries
Assay from Suppliers CofA
≥96.0% (U.S. specification)
Identification (FTIR)
Conforms

概要

N,N'-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text