Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): L04694.22
100 g, Each
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): L04694.22
100 g, Each
数量
製品番号(カタログ番号): L04694.22
または、製品番号L04694-22

化学物質識別子

CAS
110-26-9
IUPAC Name
N-[(prop-2-enamido)methyl]prop-2-enamide
Molecular Formula
C7H10N2O2
InChI Key
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES
C=CC(=O)NCNC(=O)C=C
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%
Appearance (Color)
White
Identification (FTIR)
Conforms

概要

N,N'-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.

Solubility
Soluble in water, ethanol and methanol.

Notes
Air and light sensitive. Store in a cool place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron, iron salts, copper, aluminum, brass and free radical initiators.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text