Thermo Scientific Chemicals

1,3-Diphenylpropane, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): L08849.04
2 g, Each
Thermo Scientific Chemicals

1,3-Diphenylpropane, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): L08849.04
2 g, Each
数量
製品番号(カタログ番号): L08849.04
または、製品番号L08849-04

化学物質識別子

CAS
1081-75-0
IUPAC Name
(3-phenylpropyl)benzene
Molecular Formula
C15H16
InChI Key
VEAFKIYNHVBNIP-UHFFFAOYSA-N
SMILES
C(CC1=CC=CC=C1)CC1=CC=CC=C1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.5580-1.5620 @ 20°C

概要

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text