Thermo Scientific Chemicals

1-Hydroxycyclohexyl phenyl ketone, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): L11377.36
500 g, Each
Thermo Scientific Chemicals

1-Hydroxycyclohexyl phenyl ketone, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): L11377.36
500 g, Each
数量
製品番号(カタログ番号): L11377.36
または、製品番号L11377-36

化学物質識別子

CAS
947-19-3
IUPAC Name
1-benzoylcyclohexan-1-ol
Molecular Formula
C13H16O2
InChI Key
QNODIIQQMGDSEF-UHFFFAOYSA-N
SMILES
OC1(CCCCC1)C(=O)C1=CC=CC=C1
Appearance (Color)
White
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
45.0-51.0?C

概要

1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.

Solubility
Slightly soluble in water (1108 mg/L at 25°C). Soluble in acetone, butyl acetate, methanol and toluene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text