Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylpyridine, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): L14143.03
1 g, Each
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylpyridine, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号): L14143.03
1 g, Each
数量
製品番号(カタログ番号): L14143.03
または、製品番号L14143-03

化学物質識別子

CAS
38222-83-2
IUPAC Name
2,6-di-tert-butyl-4-methylpyridine
Molecular Formula
C14H23N
InChI Key
HVHZEKKZMFRULH-UHFFFAOYSA-N
SMILES
CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
Appearance (Color)
White to pale yellow or brown
Form
Crystals or powder or crystalline powder or crystalline chunks or fused/lumpy solid
Assay (GC)
≥96.0%
Refractive Index
1.4745-1.4795 as melt

概要

2,6-Di-tert-butyl-4-methylpyridine is used for the generation of enol triflates from ketones using trifluoromethanesulfonic anhydride. It is also used in the synthesis of 1,2-dihydro-2-silanaphthalene derivatives, as base in PtCl4-catalyzed cyclization reactions of homopropargyl azide derivatives and in diastereoselective synthesis of β-thiomannopyranosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butyl-4-methylpyridine is used for the generation of enol triflates from ketones using trifluoromethanesulfonic anhydride. It is also used in the synthesis of 1,2-dihydro-2-silanaphthalene derivatives, as base in PtCl4-catalyzed cyclization reactions of homopropargyl azide derivatives and in diastereoselective synthesis of β-thiomannopyranosides.

Solubility
Sparingly soluble in water. Soluble in ethanol, acetic acid and diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to air and light.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text