Thermo Scientific Chemicals

N-Methoxymethyl-N-(trimethylsilylmethyl)benzylamine, 94%, Thermo Scientific Chemicals

製品番号(カタログ番号): L19412.03
1 g, Each
Thermo Scientific Chemicals

N-Methoxymethyl-N-(trimethylsilylmethyl)benzylamine, 94%, Thermo Scientific Chemicals

製品番号(カタログ番号): L19412.03
1 g, Each
数量
製品番号(カタログ番号): L19412.03
または、製品番号L19412-03

化学物質識別子

CAS
93102-05-7
IUPAC Name
benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine
Molecular Formula
C13H23NOSi
InChI Key
RPZAAFUKDPKTKP-UHFFFAOYSA-N
SMILES
COCN(CC1=CC=CC=C1)C[Si](C)(C)C
Appearance (Color)
Clear, colorless to pale yellow
Assay (Non-aqueous acid-base Titration)
>92.5%
Refractive Index
1.4885-1.4955 @ 20?C
Form
Liquid
Proton NMR
Conforms to structure

概要

N-Methoxymethyl-N-(trimethylsilylmethyl)benzylamine forms azomethine ylides which readily undergo [3+2] cycloaddition to α,β-unsaturated esters affording N-benzyl substituted pyrrolidines in good yields. It reacts with asymmetric 1,3-dipolar cycloadditions in the practical, large-scale synthesis of chiral pyrrolidines. It is used in the the synthesis of 3-carboxy-1-azabicyclo[2.2.1]heptane derivatives, an important class of physiologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methoxymethyl-N-(trimethylsilylmethyl)benzylamine forms azomethine ylides which readily undergo [3+2] cycloaddition to α,ß-unsaturated esters affording N-benzyl substituted pyrrolidines in good yields. It reacts with asymmetric 1,3-dipolar cycloadditions in the practical, large-scale synthesis of chiral pyrrolidines. It is used in the the synthesis of 3-carboxy-1-azabicyclo[2.2.1]heptane derivatives, an important class of physiologically active compounds.

Solubility
Soluble in chloroform, ethyl acetate.

Notes
Moisture & Light Sensitive. Store away from oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text