4-Fluoro-L-phenylalanine, 98+%, Thermo Scientific Chemicals
4-Fluoro-L-phenylalanine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-L-phenylalanine, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号) L19934.06
または、製品番号L19934-06
価格(JPY)/ Each
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1132-68-9
IUPAC Name2-amino-3-(4-fluorophenyl)propanoic acid
Molecular FormulaC9H10FNO2
InChI KeyXWHHYOYVRVGJJY-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(F)C=C1)C(O)=O
さらに表示
Appearance (Color)White to off-white to pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0% (non-U.S. specification)
Assay from Suppliers CofA≥97.0% (U.S. specification)
Optical Rotation-24° ± 1.5° (c=1, water, non-U.S. specification)
さらに表示
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

Solubility
Soluble in water and 0.5M HCl (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. R.Shiman; D.W.Gray; M.A.Hill. Regulation of rat liver phenylalanine hydroxylase. I. Kinetic properties of the enzyme's iron and enzyme reduction site. Journal of Biological Chemistry. 1994, 269,(40), 24637-24646.
  2. P.J.Hillas; P.F.Fitzpatrick. A mechanism for hydroxylation by tyrosine hydroxylase based on partitioning of substituted phenylalanines. Biochemistry (Washington). 1996, 35,(22), 6969-6975.