Methanol, Fisher Chemical™
Methanol, Fisher Chemical™
Fisher Chemical™

Methanol, Fisher Chemical™

Methanol Certified AR
製品番号(カタログ番号) M-4000-27
または、製品番号M/4000/27
価格(JPY)
-
お問い合わせください ›
Ask our AI about this Product
Methanol Certified AR
  • Methanol Certified AR
  • Plastic Bottle
  • CAS No. 67-56-1
仕様
化学物質名または材質Methanol
融点-98°C (literature)
CAS67-56-1
沸点65.4°C (literature)
数量200 L
パッケージPlastic Bottle
分子式CH4O
MDL番号MFCD00004595
InChIキーOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
IUPAC名methanol
分子量(g/mol)32.04
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

0件の結果が表示されました。 上記から特定の証明書を検索します

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection