Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-92 |
---|---|
miRBase Accession Number: | MI0000719 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAUUGCACUUGUCCCGGCCUG |
Species: | Mouse, Rat, Chicken, Oryzias latipes, Xenopus tropicalis |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000013, mir-25 |
Mature miRNA Sequence: | UAUUGCACUUGUCCCGGCCUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-92a-3p | MIMAT0000539 |
Rat | rno-miR-92a-3p | MIMAT0000816 |
Chicken | gga-miR-92-3p | MIMAT0001109 |
Oryzias latipes | ola-miR-92a | MIMAT0022534 |
Xenopus tropicalis | xtr-miR-92a | MIMAT0003580 |